siRNA Details
| Gene Symbol: Adh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012084.1 | NP_001012084.1 | 6 | 828 | ||
| XM_006233372.2 | XP_006233434.1 | 6 | 1622 | |||
| XM_006233374.1 | XP_006233436.1 | 7 | 900 | |||
| XM_006233375.2 | XP_006233437.1 | 8 | 1111 | |||
| XM_006233377.2 | XP_006233439.1 | 6 | 1070 | |||
| XM_006233378.2 | XP_006233440.1 | 7 | 980 | |||
| XM_006233379.2 | XP_006233441.1 | 7 | 977 | |||
| XM_008761506.1 | XP_008759728.1 | 5 | 942 | |||
| XM_008761507.1 | XP_008759729.1 | 7 | 3174 | |||
| XM_008761508.1 | XP_008759730.1 | 8 | 3034 | |||
| XM_008761509.1 | XP_008759731.1 | 8 | 1526 | |||
| XM_008761510.1 | XP_008759732.1 | 8 | 3302 | |||
| GenBank mRNA | BC083792.1 | - | 6 | 828 | 7106499 | |

Adh6
Genomic Map