siRNA Details
| Gene Symbol: Prkra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024780.1 | NP_001019951.1 | 8 | 1032 | ||
| GenBank mRNA | BC097446.1 | - | 8 | 1032 | 7456528 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkra| Gene Symbol: Prkra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024780.1 | NP_001019951.1 | 8 | 1032 | ||
| GenBank mRNA | BC097446.1 | - | 8 | 1032 | 7456528 | |