siRNA Details
| Gene Symbol: Pacsin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009966.2 | NP_001009966.1 | 3, 4 | 312 | ||
| XM_006234509.2 | XP_006234571.1 | 4, 5 | 511 | |||
| XM_006234510.2 | XP_006234572.1 | 4, 5 | 432 | |||
| XM_006234511.2 | XP_006234573.1 | 3, 4 | 470 | |||
| XM_008762007.1 | XP_008760229.1 | 4, 5 | 498 | |||
| GenBank mRNA | AY858802.1 | - | 2, 3 | 222 | ||
| BC100636.1 | - | 3, 4 | 312 | 5620884 | ||

Pacsin3
Genomic Map