siRNA Details
| Gene Symbol: Pacsin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009966.2 | NP_001009966.1 | 4 | 498 | ||
| XM_006234509.2 | XP_006234571.1 | 5 | 697 | |||
| XM_006234510.2 | XP_006234572.1 | 5 | 618 | |||
| XM_006234511.2 | XP_006234573.1 | 4 | 656 | |||
| XM_008762007.1 | XP_008760229.1 | 5 | 684 | |||
| GenBank mRNA | AY858802.1 | - | 3 | 408 | ||
| BC100636.1 | - | 4 | 498 | 5620884 | ||

Pacsin3
Genomic Map