siRNA Details
| Gene Symbol: Tmem127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100978.1 | NP_001094448.1 | 2 | 435 | ||
| GenBank mRNA | BC131850.1 | - | 2 | 435 | 7125680 | |
| FQ230469.1 | - | 2 | 409 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem127| Gene Symbol: Tmem127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100978.1 | NP_001094448.1 | 2 | 435 | ||
| GenBank mRNA | BC131850.1 | - | 2 | 435 | 7125680 | |
| FQ230469.1 | - | 2 | 409 | |||