siRNA Details
| Gene Symbol: Spef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039024.2 | NP_001034113.2 | 2 | 263 | ||
| XM_006235033.2 | XP_006235095.1 | 2 | 2617 | |||
| GenBank mRNA | BC099127.1 | - | 2 | 263 | 7457521 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spef1| Gene Symbol: Spef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039024.2 | NP_001034113.2 | 2 | 263 | ||
| XM_006235033.2 | XP_006235095.1 | 2 | 2617 | |||
| GenBank mRNA | BC099127.1 | - | 2 | 263 | 7457521 | |