siRNA Details
| Gene Symbol: LOC312273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107856.1 | NP_001101326.1 | 4 | 561 | ||
| GenBank mRNA | X59012.1 | - | 4 | 564 | ||
| X59013.1 | - | 4 | 564 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC312273| Gene Symbol: LOC312273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107856.1 | NP_001101326.1 | 4 | 561 | ||
| GenBank mRNA | X59012.1 | - | 4 | 564 | ||
| X59013.1 | - | 4 | 564 | |||