siRNA Details
| Gene Symbol: Actl7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025417.1 | NP_001020588.1 | 1 | 408 | ||
| GenBank mRNA | BC097412.1 | - | 1 | 408 | 7454223 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Actl7b| Gene Symbol: Actl7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025417.1 | NP_001020588.1 | 1 | 408 | ||
| GenBank mRNA | BC097412.1 | - | 1 | 408 | 7454223 | |