siRNA Details
| Gene Symbol: Phf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107995.1 | NP_001101465.1 | 1 | 112 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phf13| Gene Symbol: Phf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107995.1 | NP_001101465.1 | 1 | 112 | ||