siRNA Details
Gene Symbol: Celf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135603.1 | NP_001129075.1 | 5 | 550 | ||
XM_006240941.2 | XP_006241003.2 | 5 | 544 | |||
XM_006240942.2 | XP_006241004.2 | 5 | 517 | |||
XM_008765104.1 | XP_008763326.1 | 4 | 353 | |||
XM_008765105.1 | XP_008763327.1 | 5 | 542 | |||
XM_008765106.1 | XP_008763328.1 | 5 | 544 |
