siRNA Details
| Gene Symbol: Celf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135603.1 | NP_001129075.1 | 6 | 619 | ||
| XM_006240941.2 | XP_006241003.2 | 6 | 613 | |||
| XM_006240942.2 | XP_006241004.2 | 6 | 586 | |||
| XM_008765104.1 | XP_008763326.1 | 5 | 422 | |||
| XM_008765105.1 | XP_008763327.1 | 6 | 611 | |||
| XM_008765106.1 | XP_008763328.1 | 6 | 613 | |||

Celf5
Genomic Map