siRNA Details
| Gene Symbol: C1qtnf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012123.1 | NP_001012123.1 | 3 | 862 | ||
| GenBank mRNA | BC089992.1 | - | 3 | 862 | 7319641 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1qtnf5| Gene Symbol: C1qtnf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012123.1 | NP_001012123.1 | 3 | 862 | ||
| GenBank mRNA | BC089992.1 | - | 3 | 862 | 7319641 | |