siRNA Details
| Gene Symbol: Kdelc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025123.3 | NP_001020294.3 | 7, 8 | 1407 | ||
| GenBank mRNA | BC093594.1 | - | 7, 8 | 1407 | 7314531 | |
| FQ211817.1 | - | 6, 7 | 1111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kdelc2| Gene Symbol: Kdelc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025123.3 | NP_001020294.3 | 7, 8 | 1407 | ||
| GenBank mRNA | BC093594.1 | - | 7, 8 | 1407 | 7314531 | |
| FQ211817.1 | - | 6, 7 | 1111 | |||