siRNA Details
| Gene Symbol: Kdelc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025123.3 | NP_001020294.3 | 6 | 1205 | ||
| GenBank mRNA | BC093594.1 | - | 6 | 1205 | 7314531 | |
| BC100617.1 | - | 6 | 1204 | 7934552 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kdelc2| Gene Symbol: Kdelc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025123.3 | NP_001020294.3 | 6 | 1205 | ||
| GenBank mRNA | BC093594.1 | - | 6 | 1205 | 7314531 | |
| BC100617.1 | - | 6 | 1204 | 7934552 | ||