siRNA Details
| Gene Symbol: Loxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012125.1 | NP_001012125.1 | 4 | 1831 | ||
| GenBank mRNA | BC089224.1 | - | 4 | 1831 | 7309311 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Loxl1| Gene Symbol: Loxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012125.1 | NP_001012125.1 | 4 | 1831 | ||
| GenBank mRNA | BC089224.1 | - | 4 | 1831 | 7309311 | |