siRNA Details
| Gene Symbol: Larp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108154.1 | NP_001101624.1 | 3 | 938 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Larp6| Gene Symbol: Larp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108154.1 | NP_001101624.1 | 3 | 938 | ||