siRNA Details
Gene Symbol: RGD1562431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_237227.4 | XP_237227.3 | 1 | 1296 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1562431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_237227.4 | XP_237227.3 | 1 | 1296 |