siRNA Details
| Gene Symbol: Shd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108223.1 | NP_001101693.1 | 6, 7 | 1059 | ||
| XM_006244329.1 | XP_006244391.1 | 6, 7 | 3148 | |||
| GenBank mRNA | BC158646.1 | - | 6, 7 | 1001 | 8364383 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shd| Gene Symbol: Shd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108223.1 | NP_001101693.1 | 6, 7 | 1059 | ||
| XM_006244329.1 | XP_006244391.1 | 6, 7 | 3148 | |||
| GenBank mRNA | BC158646.1 | - | 6, 7 | 1001 | 8364383 | |