siRNA Details
| Gene Symbol: Itm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025712.1 | NP_001020883.1 | 5 | 763 | ||
| GenBank mRNA | BC099174.1 | - | 5 | 763 | 7382658 | |
| FQ217144.1 | - | 5 | 782 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itm2a| Gene Symbol: Itm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025712.1 | NP_001020883.1 | 5 | 763 | ||
| GenBank mRNA | BC099174.1 | - | 5 | 763 | 7382658 | |
| FQ217144.1 | - | 5 | 782 | |||