siRNA Details
| Gene Symbol: Itm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025712.1 | NP_001020883.1 | 1 | 200 | ||
| GenBank mRNA | BC099174.1 | - | 1 | 200 | 7382658 | |
| FQ216632.1 | - | 1 | 201 | |||
| FQ217144.1 | - | 1 | 219 | |||
| FQ224887.1 | - | 1 | 219 | |||

Itm2a
Genomic Map