siRNA Details
| Gene Symbol: LOC100360128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002730175.3 | XP_002730221.1 | 1 | 759 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100360128| Gene Symbol: LOC100360128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002730175.3 | XP_002730221.1 | 1 | 759 | ||