siRNA Details
| Gene Symbol: Lat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173840.1 | NP_776212.1 | 10, 11 | 520 | ||
| XM_006249160.2 | XP_006249222.1 | 11, 12 | 691 | |||
| XM_008769138.1 | XP_008767360.1 | 10, 11 | 973 | |||
| XM_008769139.1 | XP_008767361.1 | 9, 10 | 937 | |||
| GenBank mRNA | AY170849.1 | - | 10, 11 | 520 | ||
| FQ231408.1 | - | 8, 9 | 380 | |||
| FQ235223.1 | - | 8, 9 | 380 | |||

Lat2
Genomic Map