siRNA Details
| Gene Symbol: Lat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173840.1 | NP_776212.1 | 4 | 226 | ||
| XM_006249160.2 | XP_006249222.1 | 5 | 397 | |||
| XM_008769138.1 | XP_008767360.1 | 4 | 679 | |||
| XM_008769139.1 | XP_008767361.1 | 4 | 679 | |||
| GenBank mRNA | AY170849.1 | - | 4 | 226 | ||
| FQ231408.1 | - | 2 | 86 | |||
| FQ235223.1 | - | 2 | 86 | |||

Lat2
Genomic Map