siRNA Details
Gene Symbol: Myod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176079.1 | NP_788268.1 | 3 | 1069 | ||
GenBank mRNA | BC127480.1 | - | 3 | 1049 | 8366406 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Myod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176079.1 | NP_788268.1 | 3 | 1069 | ||
GenBank mRNA | BC127480.1 | - | 3 | 1049 | 8366406 |