siRNA Details
| Gene Symbol: Ifngr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108313.1 | NP_001101783.1 | 6 | 910 | ||
| GenBank mRNA | FQ227918.1 | - | 2 | 200 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifngr2| Gene Symbol: Ifngr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108313.1 | NP_001101783.1 | 6 | 910 | ||
| GenBank mRNA | FQ227918.1 | - | 2 | 200 | ||