siRNA Details
| Gene Symbol: Ap1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108331.2 | NP_001101801.2 | 4 | 466 | ||
| XM_006249179.2 | XP_006249241.1 | 4 | 495 | |||
| GenBank mRNA | BC169070.1 | - | 4 | 410 | 5598714 | |
| FQ230081.1 | - | 3 | 330 | |||

Ap1s1
Genomic Map