siRNA Details
| Gene Symbol: Ap1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108331.2 | NP_001101801.2 | 1, 2 | 88 | ||
| XM_006249179.2 | XP_006249241.1 | 1, 2 | 117 | |||
| GenBank mRNA | BC169070.1 | - | 1, 2 | 32 | 5598714 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap1s1| Gene Symbol: Ap1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108331.2 | NP_001101801.2 | 1, 2 | 88 | ||
| XM_006249179.2 | XP_006249241.1 | 1, 2 | 117 | |||
| GenBank mRNA | BC169070.1 | - | 1, 2 | 32 | 5598714 | |