siRNA Details
| Gene Symbol: Tacc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004424.1 | NP_001004424.1 | 2 | 221 | ||
| XM_006251329.2 | XP_006251391.1 | 2 | 241 | |||
| XM_006251330.2 | XP_006251392.1 | 2 | 244 | |||
| XM_006251331.2 | XP_006251393.1 | 2 | 236 | |||

Tacc3
Genomic Map