siRNA Details
| Gene Symbol: Smim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_341397.6 | XP_341398.4 | 1 | 129 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim4| Gene Symbol: Smim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_341397.6 | XP_341398.4 | 1 | 129 | ||