siRNA Details
| Gene Symbol: Pqlc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013189.1 | NP_001013207.1 | 1 | 291 | ||
| XM_006254952.2 | XP_006255014.1 | 2 | 583 | |||
| XM_006254953.2 | XP_006255015.1 | 2 | 976 | |||
| XM_006254954.1 | XP_006255016.1 | 1 | 751 | |||
| XM_006254955.2 | XP_006255017.1 | 2 | 582 | |||
| XM_006254956.2 | XP_006255018.1 | 2 | 973 | |||
| XM_008772155.1 | XP_008770377.1 | 1 | 751 | |||
| XM_008772156.1 | XP_008770378.1 | 1 | 751 | |||
| GenBank mRNA | BC079072.1 | - | 1 | 749 | ||
| BC088186.1 | - | 1 | 291 | 7369419 | ||

Pqlc1
Genomic Map