siRNA Details
| Gene Symbol: LOC102555567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255677.2 | XP_006255739.2 | 5 | 721 | ||
| Gene Symbol: Nudt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108450.1 | NP_001101920.1 | 5 | 2360 | ||

LOC102555567
Genomic Map