siRNA Details
| Gene Symbol: Scgb1b30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100859.1 | NP_001094329.1 | 2 | 174 | ||
| XM_008759171.1 | XP_008757393.1 | 2 | 304 | |||
| GenBank mRNA | AY871780.1 | - | 1 | 107 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb1b30| Gene Symbol: Scgb1b30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100859.1 | NP_001094329.1 | 2 | 174 | ||
| XM_008759171.1 | XP_008757393.1 | 2 | 304 | |||
| GenBank mRNA | AY871780.1 | - | 1 | 107 | ||