siRNA Details
| Gene Symbol: Fam71e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229179.2 | XP_006229241.1 | 3, 4 | 485 | ||
| XM_006229180.2 | XP_006229242.1 | 3, 4 | 485 | |||
| XM_006229182.2 | XP_006229244.1 | 3, 4 | 485 | |||
| XM_006229183.2 | XP_006229245.1 | 2, 3 | 366 | |||
| XM_008759449.1 | XP_008757671.1 | 3, 4 | 485 | |||
| XM_008759450.1 | XP_008757672.1 | 3, 4 | 542 | |||
| XM_341847.7 | XP_341848.3 | 3, 4 | 485 | |||

Fam71e1
Genomic Map