siRNA Details
| Gene Symbol: LOC102548136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC161889.1 | - | 1 | 370 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102548136| Gene Symbol: LOC102548136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC161889.1 | - | 1 | 370 | ||