siRNA Details
| Gene Symbol: Lhpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009706.1 | NP_001009706.1 | 3 | 335 | ||
| XM_008759911.1 | XP_008758133.1 | 3 | 362 | |||
| XM_008759912.1 | XP_008758134.1 | 3 | 1720 | |||
| GenBank mRNA | BC088448.1 | - | 3 | 335 | 7126522 | |

Lhpp
Genomic Map