siRNA Details
| Gene Symbol: Lrrc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143756.1 | NP_001137228.1 | 9 | 1359 | ||
| XM_006230495.2 | XP_006230557.1 | 9 | 1433 | |||
| XM_008760008.1 | XP_008758230.1 | 9 | 2261 | |||
| GenBank mRNA | BC158629.1 | - | 8 | 1257 | 7456037 | |
| CK597793.1 | - | 3 | 534 | |||

Lrrc27
Genomic Map