siRNA Details
| Gene Symbol: Tmem252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007738.1 | NP_001007739.1 | 1 | 105 | ||
| GenBank mRNA | BC079347.1 | - | 1 | 105 | 7107289 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem252| Gene Symbol: Tmem252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007738.1 | NP_001007739.1 | 1 | 105 | ||
| GenBank mRNA | BC079347.1 | - | 1 | 105 | 7107289 | |