siRNA Details
| Gene Symbol: Zdhhc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037652.1 | NP_001032741.1 | 1339 | |||
| GenBank mRNA | AY886524.1 | - | 7, 8 | 1220 | ||
| BC107928.1 | - | 1339 | 7130799 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zdhhc6| Gene Symbol: Zdhhc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037652.1 | NP_001032741.1 | 1339 | |||
| GenBank mRNA | AY886524.1 | - | 7, 8 | 1220 | ||
| BC107928.1 | - | 1339 | 7130799 | |||