siRNA Details
| Gene Symbol: Nfkbil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212509.2 | NP_997674.1 | 4 | 1090 | ||
| XM_006256046.2 | XP_006256108.1 | 4 | 1025 | |||
| GenBank mRNA | BC099772.1 | - | 4 | 1034 | 7932537 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfkbil1| Gene Symbol: Nfkbil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212509.2 | NP_997674.1 | 4 | 1090 | ||
| XM_006256046.2 | XP_006256108.1 | 4 | 1025 | |||
| GenBank mRNA | BC099772.1 | - | 4 | 1034 | 7932537 | |