siRNA Details
| Gene Symbol: RGD1359508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009707.1 | NP_001009707.1 | 3 | 200 | ||
| XM_006232306.2 | XP_006232368.1 | 3 | 476 | |||
| XM_006232307.2 | XP_006232369.1 | 2 | 171 | |||
| XM_006232308.2 | XP_006232370.1 | 3 | 262 | |||
| XM_006232309.2 | XP_006232371.1 | 3 | 262 | |||
| GenBank mRNA | BC088236.1 | - | 3 | 200 | 7325191 | |

RGD1359508
Genomic Map