siRNA Details
| Gene Symbol: Slc39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134577.1 | NP_001128049.1 | 4 | 461 | ||
| XM_006232707.2 | XP_006232769.1 | 4 | 509 | |||
| GenBank mRNA | BC168649.1 | - | 4 | 461 | 7105124 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc39a1| Gene Symbol: Slc39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134577.1 | NP_001128049.1 | 4 | 461 | ||
| XM_006232707.2 | XP_006232769.1 | 4 | 509 | |||
| GenBank mRNA | BC168649.1 | - | 4 | 461 | 7105124 | |