siRNA Details
| Gene Symbol: Lrit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232007.1 | XP_006232069.1 | 2 | 489 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrit3| Gene Symbol: Lrit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232007.1 | XP_006232069.1 | 2 | 489 | ||