siRNA Details
| Gene Symbol: Loh12cr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108650.2 | NP_001102120.1 | 4 | 439 | ||
| XM_008763394.1 | XP_008761616.1 | 4 | 1170 | |||
| XM_008763395.1 | XP_008761617.1 | 4 | 1170 | |||
| GenBank mRNA | BC160872.1 | - | 4 | 439 | 7365768 | |
| FQ211769.1 | - | 4 | 534 | |||

Loh12cr1
Genomic Map