siRNA Details
| Gene Symbol: Wwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024757.1 | NP_001019928.1 | 8 | 868 | ||
| XM_006237930.2 | XP_006237992.1 | 8 | 783 | |||
| XM_006237931.2 | XP_006237993.1 | 7 | 1210 | |||
| XM_008763590.1 | XP_008761812.1 | 9 | 1081 | |||
| GenBank mRNA | BC097386.1 | - | 8 | 868 | 7446512 | |

Wwp1
Genomic Map