siRNA Details
| Gene Symbol: Ccl27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108660.1 | NP_001102130.1 | 4 | 545 | ||
| XM_006238073.2 | XP_006238135.1 | 7 | 498 | |||
| XM_006238074.2 | XP_006238136.1 | 7 | 495 | |||
| XM_006238076.1 | XP_006238138.1 | 5 | 324 | |||
| XM_006238077.2 | XP_006238139.1 | 5 | 337 | |||
| XM_006238079.2 | XP_006238141.1 | 5 | 293 | |||
| XM_006238081.1 | XP_006238143.1 | 2 | 990 | |||
| XM_008763627.1 | XP_008761849.1 | 7 | 453 | |||
| GenBank mRNA | FQ228666.1 | - | 2 | 125 | ||

Ccl27
Genomic Map