siRNA Details
Gene Symbol: Letmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122781.3 | NP_001116253.3 | 7, 8 | 943 | ||
XM_006242354.2 | XP_006242416.1 | 7, 8 | 1006 | |||
GenBank mRNA | BC159426.1 | - | 7, 8 | 924 | 9034024 | |
DN933141.1 | - | 1, 2 | 165 |
