siRNA Details
| Gene Symbol: Letmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122781.3 | NP_001116253.3 | 7 | 865 | ||
| XM_006242354.2 | XP_006242416.1 | 7 | 928 | |||
| GenBank mRNA | BC159426.1 | - | 7 | 846 | 9034024 | |
| DN933141.1 | - | 1 | 87 | |||

Letmd1
Genomic Map