siRNA Details
Gene Symbol: Pate4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108757.1 | NP_001102227.1 | 2 | 191 | ||
XM_008766061.1 | XP_008764283.1 | 1 | 325 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pate4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108757.1 | NP_001102227.1 | 2 | 191 | ||
XM_008766061.1 | XP_008764283.1 | 1 | 325 |