siRNA Details
| Gene Symbol: Nmnat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013224.1 | NP_001013242.1 | 5 | 584 | ||
| XM_006243608.2 | XP_006243670.1 | 5 | 928 | |||
| XM_006243609.2 | XP_006243671.1 | 5 | 932 | |||
| XM_006243610.2 | XP_006243672.1 | 5 | 548 | |||
| XM_008766541.1 | XP_008764763.1 | 3 | 670 | |||
| GenBank mRNA | BC083725.1 | - | 5 | 584 | 7191168 | |

Nmnat3
Genomic Map