siRNA Details
| Gene Symbol: Fbxo36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108804.1 | NP_001102274.1 | 4 | 441 | ||
| XM_008767272.1 | XP_008765494.1 | 4 | 800 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo36| Gene Symbol: Fbxo36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108804.1 | NP_001102274.1 | 4 | 441 | ||
| XM_008767272.1 | XP_008765494.1 | 4 | 800 | |||